SPE Plates, 96-well, NBE MAESTRO
Katalog # TCAT613-0051NBE
Leverantör: Tecan
Specifikationer
- Ant. brunnar:96
- Beskrivning:Plate NBE MAESTRO 1 ml; 10 µm
- Phase:MAESTRO
- Fyllning:5 mg
- Förp.:1
Specifikationer
Om denna produkt
Silica-based sorbent with C18 bonding in proprietary coverage.
- Reversed-phased
- Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.