Anti-ICAM-2/CD102 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-32236
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:ICAM-2/CD102
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:3384
- Antigen synonyms:CD102 antigen|ICAM-2|CD102|intercellular adhesion molecule 2
- Storage buffer:Tris-Citrate/Phosphate (pH 7,0 - 8,0)
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Intercellular Adhesion Molecule 2 using the numbering given in entry NP_001093256.1 (GeneID 3384).
- Purification:Immunogen affinity purified
- Pk:100 µl
Specifications
About this item
The ICAM-2/CD102 antibody from Novus Biologicals is a rabbit polyclonal antibody to ICAM-2/CD102. This antibody reacts with human. The ICAM-2/CD102 antibody has been validated for the following applications: western blot.
Type: Primary
Antigen: ICAM-2/CD102
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human