Sodium nitrite ≥98.0% ACS, Reag. USP, Reag. Ph. Eur analytical reagent
Leverantör: PanReac AppliChem
Certifikat
Om denna produkt
CAS-nummer:7632-00-0
:
Formel:NaNO₂
MDL-nummer:MFCD00011118
:69 g/mol
Smält Pt:271 °C
Förvaringstemperatur:Ambient
EINECS:231-555-9
FN:1500
ADR:5.1 (6.1),III
:GHS03
:GHS06
:GHS09
Specifikationstestresultat
- Assay (Perm.):min. 98.0%
- Insoluble matter in H2O:max. 0.003 %
- Chloride (Cl):max. 0.002%
- Sulfate (SO4):max. 0.005%
- As:max. 0.00004 %
- Heavy metals (as Pb):max. 0.001%
- Al:max. 5 ppm
- Au:max. 5 ppm
- B:max. 5 ppm
- Ba:max. 5 ppm
- Be:max. 5 ppm
- Bi:max. 5 ppm
- Ca:max. 25 ppm
- Cd:max. 5 ppm
- Co:max. 5 ppm
- Cr:max. 5 ppm
- Cu:max. 10 ppm
- Fe:max. 10 ppm
- Ga:max. 5 ppm
- Ge:max. 5 ppm
- Hg:max. 5 ppm
- K:max. 50 ppm
- Li:max. 5 ppm
- MG:max. 25 ppm
- Mn:max. 5 ppm
- Mo:max. 5 ppm
- Ni:max. 10 ppm
- PB:max. 10 ppm
- Sb:max. 5 ppm
- Si:max. 5 ppm
- Sn:max. 5 ppm
- Sr:max. 5 ppm
- Ti:max. 5 ppm
- Tl:max. 5 ppm
- V:max. 5 ppm
- Zn:max. 10 ppm