Magnetic Stirring Bars
Supplier: KARTELL
Certificates
About this item
Economical stirring bars designed for a wide variety of applications.
- Stronger mixing action at lower speeds
- Can be used as an addition to any standard magnetic stir bar range, creating a viable alternative
- The sizes shown below are nominal, with ±5% tolerance in length and ±10% in diameter
Suitable for foodstuff.