Microfibre Cloths
Leverantör: Rubbermaid Commercial Products
Certifikat
Om denna produkt
Reduce the spread of germs with quality microfibre cloths that provide excellent cleaning performance.
- Versatile microfibre cloths keep all types of work surfaces clean
- Cloths remove 99,7% or more of tested virus and bacteria with water only
- Superior cleaning performance and germ removal compared to traditional cloths
- Facilitate colour-coding implementation by assigning a coloured pad to a specific area
The Rubbermaid Commercial Microfibre Cloths are durable up to 200 commercial launderings and 100 launderings with bleach.