BASE Footwear Diving Safety Shoes, Lace-Up, Low, S3, SRC
PRTWB0605BKG36
: Portwest
- Shoe size:36
- Colour:Black/grey
- Pk:1 PAIR
Diving safety shoes feature a water-resistant, full grain leather upper, SmellStop anti-odour and anti-bacterial lining, Fresh’n Flex midsole, SlimCap protective toe cap, and anti-fatigue AirTech + TPU-Skin sole.
- Anti-static footwear
- Water-resistant upper to prevent water penetration
- Non-magnetic construction; nickel- and ferrous-free
- 200 joules resistant toe-cap
- Puncture-resistant sole
- Shock absorption in the heel region
- Hydrocarbons-resistant
Anti-fatigue footwear collection offers an answer to the demand for lightweight design. The orthopaedic, made to measure footbed complies with European norms DGUV112-191.
: UNI EN ISO 20345:2012 S3 SRC.