Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Itraconazole
Itraconazole, micronised USP
Itraconazole, micronised USP
Itraconazole, micronised USP
Supplier:
Spectrum Chemical
Certificates
Expand image
Itraconazole, micronised USP
Supplier:
Spectrum Chemical
About this item
CAS Number:
84625-61-6
:
Formula:
C₃₅H₃₈Cl₂N₈O₄
MDL Number:
MFCD00870168
:
705.64 g/mol
Storage Temperature:
Ambient