Order Entry
Sweden
ContactUsLinkComponent
 
Allura red AC
Catalog # SPCMFD140-100GM
Supplier:  Spectrum Chemical
CAS Number:  
Allura red AC
Catalog # SPCMFD140-100GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Identification:
    To Pass Test

Specification Test Results

About this item

 :
 :496.42 g/mol
Storage Temperature:Ambient
CAS Number:25956-17-6
Formula:C₁₈H₁₄N₂O₈S₂·2Na
MDL Number:MFCD00059526