Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethylhexyl salicylate
2-Ethylhexyl salicylate ≥95% USP
2-Ethylhexyl salicylate ≥95% USP
2-Ethylhexyl salicylate ≥95% USP
SPCMO1236-100ML
:
Spectrum Chemical
:
2-Ethylhexyl salicylate ≥95% USP
SPCMO1236-100ML
:
Spectrum Chemical
:
Pack type:
Glass, amber
Pk:
100 mL
Seq:
2440
Assay:
95% Min.
:
:
250.34 g/mol
:
Ambient
:
204-263-4
:
118-60-5
:
C₁₅H₂₂O₃
:
MFCD00053300