Order Entry
Sweden
ContactUsLinkComponent
Aminoguanidine hydrochloride
Aminoguanidine hydrochloride
Catalog # SPCMA3814-40KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Aminoguanidine hydrochloride
Catalog # SPCMA3814-40KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    40 kg
  • Seq:
    4367

Specifications

About this item

 :
 :110.55 g/mol
Storage Temperature:Ambient
CAS Number:1937-19-5
Formula:CH₇ClN₄
MDL Number:MFCD00039074