Order Entry
Sweden
ContactUsLinkComponent
D-(-)-N-Methylglucamine ≥99.0% USP
D-(-)-N-Methylglucamine ≥99.0% USP
Supplier:  Spectrum Chemical
Certificates
D-(-)-N-Methylglucamine ≥99.0% USP
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Dried Basis):
    99.0 - 100.5%
  • Completeness and Color of Solution:
    To Pass Test
  • Melting Range:
    128 - 132 °C

Specification Test Results