Benzyl salicylate ≥98%
Catalog # SPCMB2414-5KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Assay:98% Min.
Specification Test Results
About this item
:
:228.25 g/mol
Merck Index:13,01146
Boiling Pt:208 °C (1013 hPa)
Melting Pt:23.9 °C
Density:1,176
Storage Temperature:Ambient
EINECS:204-262-9
CAS Number:118-58-1
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00020034