Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gum tragacanth
Gum tragacanth, powder NF
Gum tragacanth, powder NF
Gum tragacanth, powder NF
Supplier:
Spectrum Chemical
Certificates
Expand image
Gum tragacanth, powder NF
Supplier:
Spectrum Chemical
Certificates
About this item
CAS Number:
9000-65-1
:
MDL Number:
MFCD00131255
Storage Temperature:
Ambient
EINECS:
232-552-5
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
SPCMTR105-500GM_TDS_EN
Documentation