Order Entry
Sweden
ContactUsLinkComponent
Benzyltriethylammonium chloride
Benzyltriethylammonium chloride
Catalog # SPCMB1083-45KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Benzyltriethylammonium chloride
Catalog # SPCMB1083-45KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    45 kg
  • Seq:
    4388

Specifications

About this item

 :
 :227.78 g/mol
Melting Pt:190…192 °C
Flash Pt:>100 °C
EINECS:200-270-1
 :GHS07
Storage Temperature:Ambient
CAS Number:56-37-1
Formula:C₁₃H₂₂ClN
MDL Number:MFCD00011824