Benzyl acetate ≥98%
Catalog # SPCMB2411-200LTBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Assay:98% Min.
Specification Test Results
About this item
:
:150.18 g/mol
Merck Index:13,01125
Boiling Pt:206 °C (1013 hPa)
Melting Pt:–51 °C
Density:1,04
Flash Pt:95 °C
Storage Temperature:Ambient
EINECS:205-399-7
CAS Number:140-11-4
Formula:C₉H₁₀O₂
MDL Number:MFCD00008712