Glycerine (natural) ≥99.0% USP, Ph. Eur., BP
Supplier: Spectrum Chemical
New Product
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Density:1,264
Storage Temperature:Ambient
Specification Test Results
- Assay (Anhydrous Basis):99.0 - 101.0%
- Specific Gravity @ 25°C:1.249 min.
- Appearance of Solution:To Pass Test
- Refractive Index:1.470 - 1.475
- Identification:To Pass Test