Specifications
- Antibody type:Primary
- Antigen name:C4 binding protein
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:1-100/252
- Form:Liquid
- Antigen synonyms:PRP|OTTHUMP00000034375|C4b-binding protein beta chain|OTTHUMP00000034288|OTTHUMP00000034289|OTTHUMP0000003474|Complement component 4-binding protein beta|Complement component 4 binding protein beta chain|C4BP|Complement component 4 binding protein beta|C4b binding protein beta chain|C4BPB_HUMAN|Proline rich protein|C4BPB
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:26 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C4 binding protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C4BPB is a member of a superfamily of proteins composed predominantly of tandemly arrayed short consensus repeats of approximately 60 amino acids. A single, unique beta-chain encoded by this gene assembles with seven identical alpha-chains into the predominant isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. C4b-binding protein has a regulatory role in the coagulation system also, mediated through the beta-chain binding of protein S, a vitamin K-dependent protein that serves as a cofactor of activated protein C. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Alternative splicing gives rise to multiple transcript variants
Type: Primary
Antigen: C4 binding protein
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 1-100/252
Host: Rabbit
Isotype: IgG
Reactivity: Human