Anti-DNA polymerase epsilon subunit 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-14356R-A750
: Bioss
- Antibody type:Primary
- Antigen name:DNA polymerase epsilon subunit 2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Epitope:101-200/527
- Form:Liquid
- Gene ID:5427
- Antigen synonyms:DNA polymerase epsilon subunit 2|DNA polymerase II subunit 2|DPE2|POLE2|DNA polymerase epsilon subunit B
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:59 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DNA Polymerase epsilon subunit 2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Participates in DNA repair and in chromosomal DNA replication.
Type: Primary
Antigen: DNA polymerase epsilon subunit 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/527
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse