Anti-CRNKL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-14065R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CRNKL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:201-300/848
- Form:Liquid
- Gene ID:51340
- Antigen synonyms:CRN|Crooked neck-like protein 1|HCRN|Clf1|Crooked neck homolog|CLF|SYF3|CRNKL1|CGI-201|MSTP021
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:100 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CRNKL1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Involved in pre-mRNA splicing process.
Type: Primary
Antigen: CRNKL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/848
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat