Anti-GEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-13334R-A750
: Bioss
- Antibody type:Primary
- Antigen name:GEN1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/908
- Form:Liquid
- Antigen synonyms:GEN_HUMAN|Gen|Gen homolog 1 endonuclease|FLJ40869|DKFZp781F0986|Flap endonuclease GEN homolog 1|GEN 1|GEN1|Gen homolog 1 endonuclease Drosophila
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:103 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GEN1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Endonuclease which cleaves flap structures at the junction between single-stranded DNA and double-stranded DNA. Specific for 5'-overhanging flap structures in which the 5'-upstream of the flap is completely double-stranded. Prefers the blocked-flap structures similar to those occurring at replication forks, in which the 5' single-strand overhang of the flap is double-stranded.
Type: Primary
Antigen: GEN1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/908
Host: Rabbit
Isotype: IgG
Reactivity: