Order Entry
Sweden
ContactUsLinkComponent
Anti-GEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-GEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13334R-A680
 :  Bioss
Anti-GEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13334R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    GEN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    51-150/908
  • Form:
    Liquid
  • Antigen synonyms:
    GEN_HUMAN|Gen|Gen homolog 1 endonuclease|FLJ40869|DKFZp781F0986|Flap endonuclease GEN homolog 1|GEN 1|GEN1|Gen homolog 1 endonuclease Drosophila
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    103 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human GEN1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Endonuclease which cleaves flap structures at the junction between single-stranded DNA and double-stranded DNA. Specific for 5'-overhanging flap structures in which the 5'-upstream of the flap is completely double-stranded. Prefers the blocked-flap structures similar to those occurring at replication forks, in which the 5' single-strand overhang of the flap is double-stranded.

Type: Primary
Antigen: GEN1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/908
Host: Rabbit
Isotype: IgG
Reactivity: