Anti-CMTM8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-8024R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CMTM8
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:121-173/173
- Form:Liquid
- Gene ID:152189
- Antigen synonyms:CKLF like MARVEL transmembrane domain containing 8|CKLFSF8 V2|Chemokine like factor super family member 8|CMTM 8|CKLF8_HUMAN|Cklfsf8|CMTM8|CMTM-8
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CKLFSF8 / CMTM8
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown.
Type: Primary
Antigen: CMTM8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-173/173
Host: Rabbit
Isotype: IgG
Reactivity: Mouse