Anti-MND1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7799R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MND1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:1-100/205
- Form:Liquid
- Gene ID:84057
- Antigen synonyms:RGD1309007|Meiotic nuclear division 1, S. cerevisiae. homolog of antibody|Meiotic nuclear divisions 1 homolog S. cerevisiae|MND1|Homolog of yeast MND1|AI591601|MGC134110|GAJ protein|Meiotic nuclear divisions 1 homolog|2610034E18Rik|Meiotic nuclear division protein 1 homolog
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:24 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GAJ/MND1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The product of the GAJ gene associates with HOP2 to form a stable heterodimeric complex that binds DNA and stimulates the recombinase activity of RAD51 and DMC1. Both the MND1 and HOP2 genes are indispensable for meiotic recombination.
Type: Primary
Antigen: MND1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/205
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse