Anti-TPST1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6850R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TPST1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:165-260/370
- Form:Liquid
- Gene ID:8460
- Antigen synonyms:TPST1_HUMAN|Protein tyrosine sulfotransferase 1|Protein-tyrosine sulfotransferase 1|TPST1|TPST-1|Tyrosylprotein sulfotransferase 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TPST1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Catalyzes the O-sulfation of tyrosine residues within acidic motifs of polypeptides.
Type: Primary
Antigen: TPST1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 165-260/370
Host: Rabbit
Isotype: IgG
Reactivity: Mouse