Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Fascin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6848R-A750
 :  Bioss
Anti-Fascin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6848R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Fascin
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    401-493/493
  • Form:
    Liquid
  • Gene ID:
    6624
  • Antigen synonyms:
    Fascin homolog 1 actin bundling protein Strongylocentrotus purpuratus|Singed drosophila homolog like|Fscin1|Fascin|Singed like protein|HSN|SNL|p55|FAN 1|Fascin1|Singed like fascin homolog sea urchin Drosophila|FLJ38511|55 kDa actin-bundling protein|FSCN1_HUMAN|FSCN 1|FSCN1|Singed like fascin homolog sea urchin|Singed-like protein|Strongylocentrotus purpuratus|Singed Drosophila like sea urchin fascin homolog like|Actin bundling protein|Fascin homolog 1|FAN1|55 kDa actin bundling protein|Fascin 1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    55 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Fascin
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localised to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predominantly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However, Reed Sternberg cells in Hodgkin?s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.

Type: Primary
Antigen: Fascin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-493/493
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Others