Anti-E1 Ubiquitin Activating Enzyme Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6835R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:E1 Ubiquitin Activating Enzyme
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Epitope:961-1058/1058
- Form:Liquid
- Gene ID:7317
- Antigen synonyms:A1ST|Uba1|A1S9|UBE1X|GXP1|UBE 1 |A1S9T|UBA1_HUMAN|UBE1|GXP 1|A1S9 protein|UBE 1X|A1S9T and BN75 temperature sensitivity complementing|Ubiquitin activating enzyme E1|MGC4781|Ubiquitin-like modier-activating enzyme 1|Protein A1S9|Ubiquitin-activating enzyme E1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:118 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human UBE1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternatively spliced transcript variants that encode the same protein have been described.
Type: Primary
Antigen: E1 Ubiquitin Activating Enzyme
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 961-1058/1058
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat