Anti-HNRPC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6459R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HNRPC
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:21-120/306
- Form:Liquid
- Gene ID:3183
- Antigen synonyms:HNRNP|Heterogeneous nuclear ribonucleoproteins C1/C2|Heterogeneous nuclear ribonucleoprotein C|SNRPC|hnRNPC|hnRNP C1 / hnRNP C2|Nuclear ribonucleoprotein particle C2 protein|HNRPC|hnRNP C1/C2|Nuclear ribonucleoprotein particle C1 protein|HNRPC_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:34 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HNRPC
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Binds pre-mRNA and nucleates the assembly of 40S hnRNP particles. Single HNRNPC tetramers bind 230-240 nucleotides. Trimers of HNRNPC tetramers bind 700 nucleotides. May play a role in the early steps of spliceosome assembly and pre-mRNA splicing. Interacts with poly-U tracts in the 3'-UTR or 5'-UTR of mRNA and modulates the stability and the level of translation of bound mRNA molecules.
Type: Primary
Antigen: HNRPC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/306
Host: Rabbit
Isotype: IgG
Reactivity: Rat