Anti-CADM3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6027R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CADM3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:101-200/398
- Form:Liquid
- Gene ID:57863
- Antigen synonyms:Synaptic cell adhesion molecule 3|Nectin-like protein 1|CADM3|synCAM3|Immunoglobulin superfamily member 4B|UNQ225/PRO258|TSLL1|Brain immunoglobulin receptor|TSLC1-like protein 1|IGSF4B|Necl-1|NECL1|BIgR|Cell adhesion molecule 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CADM3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Involved in the cell-cell adhesion. Has both calcium-independent homophilic cell-cell adhesion activity and calcium-independent heterophilic cell-cell adhesion activity with IGSF4, PVRL1 and PVRL3. Interaction with EPB41L1 may regulate structure or function of cell-cell junctions (By similarity).
Type: Primary
Antigen: CADM3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/398
Host: Rabbit
Isotype: IgG
Reactivity: Mouse