Anti-K7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-12388R-A680
: Bioss
- Antibody type:Primary
- Antigen name:K7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:751-850/1343
- Form:Liquid
- Gene ID:374654
- Antigen synonyms:k7|Kinesin-like protein k7|kinesin family member 7|EQYK340|kinesin like protein K7|K7_HUMAN|UNQ340
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:150 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human KIF7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
KIF7 is a 1343 amino acid protein expressed in embryonic stem cells, melanotic melanoma and Jurkat T-cells. KIF7 is a member of the KIF27 subfamily of the kinesin-like protein family and contains one kinesin-motor domain. It is suggested that KIF7 may participate in the Hedgehog (Hh) Signalling pathway by regulating the proteolysis and stability of GLI transcription factors. Hedgehog (Hh) Signalling plays a critical role in embryonic development.
Type: Primary
Antigen: K7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 751-850/1343
Host: Rabbit
Isotype: IgG
Reactivity: Mouse