Anti-EDA/Ectodermal dysplasia protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-12347R-A680
: Bioss
- Antibody type:Primary
- Antigen name:EDA/Ectodermal dysplasia protein
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:101-200/391
- Form:Liquid
- Gene ID:1896
- Antigen synonyms:TNLG7C|EDA-A2|EDA1|EDA protein|XHED|Ectodysplasin-A|ECTD1|EDA2|STHAGX1|Ectodermal dysplasia protein|EDA-A1|ODT1|ED1-A2|EDA|XLHED|ED1|HED|ED1-A1|HED1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Ectodysplasin-A, membrane form/secreted form
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Seems to be involved in epithelial-mesenchymal Signalling during morphogenesis of ectodermal organs. Isoform 1 binds only to the receptor EDAR, while isoform 3 binds exclusively to the receptor XEDAR.
Type: Primary
Antigen: EDA/Ectodermal dysplasia protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/391
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse