Order Entry
Sweden
ContactUsLinkComponent
Anti-ZNF740 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-ZNF740 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12227R-A750
Supplier:  Bioss
Anti-ZNF740 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12227R-A750
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ZNF740
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 750
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    101-193/193
  • Form:
    Liquid
  • Antigen synonymer:
    Zinc finger protein 740|ZN740_HUMAN|MGC61706|OriLyt TD-element-binding protein 7|TB7|Zfp740|ZNF740
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    22 kDa
  • Förvaringstemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from Human ZNF740
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

Specifications

About this item

May be involved in transcriptional regulation.

Type: Primary
Antigen: ZNF740
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-193/193
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse