Specifications
- Antibody type:Primary
- Antigen name:RECQL5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:1-100/991
- Form:Liquid
- Antigen synonyms:Recql5b|RecQ5|DNA helicase, RecQ like type 5|Recq5b|RecQ protein-like 5, isoform CRA_b|ATP-dependent DNA helicase Q5|RecQ helicase protein-like 5 beta|RecQ protein-like 5|DNA helicase, RECQ-like, type 5
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:109 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RECQL5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
RecQ helicases are implicated in DNA recombination and repair helicases and are critical in genome maintenance. RecQL5 is one of five known RecQ homologues in humans, and defects in three of these (BLM, WRN and RecQ4) are associated with autosomal recessive disorders that involve genomic instability and a predisposition to cancer. The exact role of ReqL5 is unclear. It has been shown in mouse embryonic stem (ES) cells that mutations in the Recql5 genes result in a significant increase in the frequency of sister chromatid exchange (SCE), whereas deleting Recql5 lead to an even higher frequency of SCE. There are three different RecQL5 isoforms - alpha, beta and gamma, produced by alternative splicing.
Type: Primary
Antigen: RECQL5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/991
Host: Rabbit
Isotype: IgG
Reactivity: