Anti-HHV DNA polymerase catalytic subunit Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10489R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HHV DNA polymerase catalytic subunit
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Epitope:1131-1235/1235
- Form:Liquid
- Antigen synonyms:UL30|DNA polymerase catalytic subunit|DPOL_HHV11|HHV1gp046
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:136 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Human herpesvirus 1 DNA polymerase catalytic subunit
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Replicates viral genomic DNA. The replication complex is composed of six viral proteins: the DNA polymerase, processivity factor, primase, primase-associated factor, helicase, and ssDNA-binding protein. Additionally, the polymerase contains an intrinsic ribonuclease H (RNase H) activity that specifically degrades RNA/DNA heteroduplexes or duplex DNA substrates in the 5' to 3' direction. Therefore, it can catalyse the excision of the RNA primers that initiate the synthesis of Okazaki fragments at a replication fork during viral DNA replication.
Type: Primary
Antigen: HHV DNA polymerase catalytic subunit
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1131-1235/1235
Host: Rabbit
Isotype: IgG
Reactivity: Virus