Order Entry
Sweden
ContactUsLinkComponent
Anti-MCM3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-MCM3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1982R-A750
Supplier:  Bioss
Anti-MCM3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1982R-A750
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    MCM3
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 750
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Epitop:
    251-350/808
  • Form:
    Liquid
  • Gen-ID:
    4172
  • Antigen synonymer:
    P1 Protein|MGC1157|P1.h|MCM3 minichromosome maintenance deficient 3|HCC5|Cervical cancer proto oncogene 5|Minichromosome maintenance deficient 3|HCC 5|hRlf beta subunit|RLF subunit beta|Replication licensing factor beta subunit|Human cervical cancer proto oncogene 5|minichromosome maintenance deficient 3|DNA replication licensing factor MCM3|DNA polymerase alpha holoenzyme associated P1|RLFB.|MCM 3|DNA polymerase alpha holoenzyme associated protein P1|P102 protein|DNA replication factor MCM3|Minichromosome maintenance protein 3|P1 h|P1 MCM3|RLF beta subunit
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    89 kDa
  • Förvaringstemperatur:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human MCM3
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

Specifications

About this item

MCM3 is one of the highly conserved mini-chromosome maintenance proteins that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.

Type: Primary
Antigen: MCM3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/808
Host: Rabbit
Isotype: IgG
Reactivity: