Anti-UCP-2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-1926R-A750
: Bioss
- Antibody type:Primary
- Antigen name:UCP-2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:201-309/309
- Form:Liquid
- Gene ID:22228
- Antigen synonyms:Mitochondrial uncoupling protein 2|Ucp2|UCP 2|UCPH|Solute carrier family 25 member 8|Slc25a8
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:34 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from Mouse UCP-2 (Mitochondrial intermembrane space domain)
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
UCP are mitochondrial transporter proteins that create proton leaks across the inner mitochondrial membrane, thus uncoupling oxidative phosphorylation from ATP synthesis. As a result, energy is dissipated in the form of heat (By similarity).
Type: Primary
Antigen: UCP-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-309/309
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat