Anti-SLC6A4/5-HTT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1893R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SLC6A4/5-HTT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Epitope:401-500/630
- Form:Liquid
- Gene ID:6532
- Antigen synonyms:Solute carrier family 6 member 4|HTT|Sodium-dependent serotonin transporter|5HTT|5-HTT|SERT|5HT transporter|hSERT|SLC6A4|OCD1|SERT1|5-HTTLPR
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:70 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human 5-HTT
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Serotonin transporter whose primary function in the central nervous system involves the regulation of serotonergic signaling via transport of serotonin molecules from the synaptic cleft back into the pre-synaptic terminal for re-utilisation. Plays a key role in mediating regulation of the availability of serotonin to other receptors of serotonergic systems. Terminates the action of serotonin and recycles it in a sodium-dependent manner.
Type: Primary
Antigen: SLC6A4/5-HTT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-500/630
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken