Order Entry
Sweden
ContactUsLinkComponent
Anti-M-RAS/Mras Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-M-RAS/Mras Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1882R-A680
Supplier:  Bioss
Anti-M-RAS/Mras Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1882R-A680
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    M-RAS/Mras
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 680
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Rat
  • Western blot:
    Yes
  • Epitop:
    105-208/208
  • Form:
    Liquid
  • Gen-ID:
    22808
  • Antigen synonymer:
    XRas.|Ras related protein RRas3|Ras related protein MRas|RRas3|M ras|Muscle Ras oncogene homologue|muscle RAS oncogene homolog precursor|R ras3|Muscle and microspikes Ras|R-ras3
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    23 kDa
  • Förvaringstemperatur:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human MRAS
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

Specifications

About this item

MRas is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. They are oncogenically activated in a significant fraction of tumors. MRas participates in reorganisation of actin cytoskeleton. It is also involved in focal adhesion and can activate MAPK signalling.

Type: Primary
Antigen: M-RAS/Mras
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 105-208/208
Host: Rabbit
Isotype: IgG
Reactivity: Rat