Anti-CNR-2/PCDHA6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-1711R-A750
: Bioss
- Antibody type:Primary
- Antigen name:CNR-2/PCDHA6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:881-950/950
- Form:Liquid
- Gene ID:56142
- Antigen synonyms:PCDH-alpha-6|Cadherln-related neuronal receptor2|CRNR2|PCDHA6|Cadherin related neuronal receptor 2|Protocadherin alpha 6.|KIAA0345 like 8|PCDHA-6|CRNR 2|CNR2|PCDA6_HUMAN|CNRS2|CNRN 2|CNRS 2|PCDH alpha6|Protocadherin alpha-6|PCDHA 6|CNRN2|CNR 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:100 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CNR-2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
PCDHA6 (Protocadherin alpha 6) is a member of the protocadherin family, a subgroup of the cadherin superfamily. Protocadherins are neural cadherin-like cell adhesion proteins that are integral membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific neuronal connections in the brain. PCDHA6 is one of fifteen tandemly arranged genes within the alpha cluster of protocadherin genes on 5q31.
Type: Primary
Antigen: CNR-2/PCDHA6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 881-950/950
Host: Rabbit
Isotype: IgG
Reactivity: Rat