Anti-Cathepsin D/CTSD Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
BOSSBS-1615R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:Cathepsin D/CTSD
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:101-200/412
- Form:Liquid
- Gene ID:1509
- Antigen synonyms:CTSD|HEL-S-130P|CPSD|CLN10|Cathepsin D
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:11/38/45 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Capsin D
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Acid protease active in intracellular protein breakdown. Involved in the pathogenesis of several diseases such as breast cancer and possibly Alzheimer disease.
Type: Primary
Antigen: Cathepsin D/CTSD
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 101-200/412
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse