Anti-AP-TNAP/ALPL Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-1535R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:AP-TNAP/ALPL
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:52-150/524
- Form:Liquid
- Gene ID:11647
- Antigen synonyms:Alpl|Akp2|Alkaline phosphatase liver/bone/kidney isozyme|Akp-2|Alkaline phosphatase, tissue-nonspecific isozyme|TNSALP|AP-TNAP|TNAP|APTNAP|Alkaline phosphatase 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from AP-TNAP/ALPL
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Alkaline phosphatase (ALP) removes phosphate groups from the 5' end of DNA and RNA, and from proteins, at high pH. Most mammals have 4 different isozymes: placental, placental like, intestinal and non tissue specific (found in liver, kidney and bone). Tissues with particularly high concentrations of ALP include the liver, bile ducts, placenta, and bone. Damaged or diseased tissue releases enzymes into the blood, so serum ALP measurements can be abnormal in many conditions, including bone disease and liver disease.
Type: Primary
Antigen: AP-TNAP/ALPL
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 52-150/524
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse