Anti-Defensin beta 2/DEFB2/HBD-2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1295R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Defensin beta 2/DEFB2/HBD-2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:21-71/71
- Form:Liquid
- Gene ID:13215
- Antigen synonyms:mBD-2|BD-2|Defensin, beta 2|Defb2|Beta-defensin 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:6 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse Defensin Beta 2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 4, an antibiotic peptide which is locally regulated by inflammation.
Type: Primary
Antigen: Defensin beta 2/DEFB2/HBD-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-71/71
Host: Rabbit
Isotype: IgG
Reactivity: Mouse