Order Entry
Sweden
ContactUsLinkComponent
 
Anti-XRCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-1218R-A750
 :  Bioss
Anti-XRCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-1218R-A750
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    XRCC1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 750
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    1-120/633
  • Form:
    Liquid
  • Gen-ID:
    7515
  • Antigen synonymer:
    RCC|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1|XRCC1
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    69 kDa
  • Förvaringstemperatur:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human XRCC1
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

 

 

Corrects defective DNA strand-break repair and sister chromatid exchange following treatment with ionizing radiation and alkylating agents.

Type: Primary
Antigen: XRCC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-120/633
Host: Rabbit
Isotype: IgG
Reactivity: Human