Anti-Synaptotagmin 1+2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-1136R-A750
: Bioss
- Antibody type:Primary
- Antigen name:synaptotagmin 1+2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:285-380/419
- Form:Liquid
- Gene ID:127833
- Antigen synonyms:SytII.|R74640|SYNAPTOTAGMIN 2|Synaptotagmin II|LOC100127887|Synaptotagmin 1, Synaptotagmin 2|SYT2|FLJ42519|SYT 2|Syt II|KIAA4194|Synaptotagmin2|SYNII|SytII.Synaptotagmin II|Syt2|mKIAA4194|SynaptotagminII|RATSYNII
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:46 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human synaptotagmin-2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).[supplied by OMIM].
Type: Primary
Antigen: synaptotagmin 1+2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 285-380/419
Host: Rabbit
Isotype: IgG
Reactivity: