Anti-PP2A alpha / beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-0029R-A680
: Bioss
- Antibody type:Primary
- Antigen name:PP2A alpha / beta
- Antigen symbol:PP2A alpha / beta
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:205-309/309
- Form:Liquid
- Gene ID:5515
- Antigen synonyms:Replication protein C|PP2Calpha|PP2Ac|PP2A-alpha|PP2CA|PPP2CA|Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform|RP-C
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:34 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PP-2A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
PP2A is the major phosphatase for microtubule-associated proteins (MAPs). PP2A can modulate the activity of phosphorylase B kinase casein kinase 2, mitogen-stimulated S6 kinase, and MAP-2 kinase. Cooperates with SGOL2 to protect centromeric cohesin from separase-mediated cleavage in oocytes specifically during meiosis I (By similarity). Can dephosphorylate SV4 large T antigen and p53/TP53. Activates RAF1 by dephosphorylating it at 'Ser-259'.
Type: Primary
Antigen: PP2A alpha / beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 205-309/309
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse