Order Entry
Sweden
ContactUsLinkComponent
Anti-ATG9A Ser735 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-ATG9A Ser735 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5199R-A750
Supplier:  Bioss
Anti-ATG9A Ser735 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5199R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATG9A Ser735
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    79065
  • Antigen synonyms:
    Autophagy-related protein 9A|APG9L1|ATG9A|MGD3208|APG9-like 1|mATG9
  • Modification:
    Phosphorylation (Ser735)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    94 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human ATG9A around the phosphorylation site of Ser735
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Involved in autophagy and cytoplasm to vacuole transport (Cvt) vesicle formation. Plays a key role in the organisation of the preautophagosomal structure/phagophore assembly site (PAS), the nucleating site for formation of the sequestering vesicle. Cycles between a juxta-nuclear trans-Golgi network compartment and late endosomes. Nutrient starvation induces accumulation on autophagosomes. Starvation-dependent trafficking requires ULK1, ATG13 and SUPT20H.

Type: Primary
Antigen: ATG9A Ser735
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat