Anti-Oct-4BOCT4B-190NT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-3816R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Oct-4BOCT4B-190NT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:201-300/360
- Form:Liquid
- Gene ID:5460
- Antigen synonyms:OCT4|Oct-4|OCT3|OTF3|Octamer-binding protein 4|OTF-3|POU domain, class 5, transcription factor 1|OTF4|Octamer-binding transcription factor 3|POU5F1|Oct-3|Octamer-binding protein 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:21/39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human OCT4B-190
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Transcription factor that binds to the octamer motif (5'-ATTTGCAT-3'). Forms a trimeric complex with SOX2 on DNA and controls the expression of a number of genes involved in embryonic development such as YES1, FGF4, UTF1 and ZFP26. Critical for early embryogenesis and for embryonic stem cell pluripotency.
Type: Primary
Antigen: Oct-4BOCT4B-190NT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/360
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat