Anti-PLAA/Phospholipase A2 activator protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-3807R-A680
: Bioss
- Antibody type:Primary
- Antigen name:PLAA/Phospholipase A2 activator protein
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-200/795
- Form:Liquid
- Gene ID:9373
- Antigen synonyms:Phospholipase A-2-activating protein|DOA1|PLA2P|PLAP|PLAA
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:87 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Phospholipase A2 activator protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Phospholipase A2 activator protein (PLAP), activates PLA2 (phospholipase A2 enzyme) and is an important mediator of eicosanoid generation. Also known as a pro-inflammatory agent, PLAP has been found in inflamed tissues and synovial fluid from patients with rheumatoid arthritis. As such, it is believed to play an important role in the regulation of inflammatory diseases. The formation of PLAP can be stimulated by IL-1 beta and TNF-alpha.
Type: Primary
Antigen: PLAA/Phospholipase A2 activator protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/795
Host: Rabbit
Isotype: IgG
Reactivity: