Specifications
- Antibody type:Primary
- Antigen name:Histone H2B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:21-126/126
- Form:Liquid
- Gene ID:3017. 8339. 8343. 8344. 8346. 8347.
- Antigen synonyms:Histone H2B type 1 N|H2B.b|HIST1H2BB|H2BFC|Histone H2B type 1 D|H2BFQ|H2BFO|HIST1H2BN|H2B histone family member S|Histone H2B type 2 E|H2BFE|H2B.q|HIST1H2BM|H2B.j|HIST2H2BE|Histone H2B|H2B GL105|H2B.1 B|H2BFJ|H2B.e|HIST1H2BL|HIST1H2BD|H2BFB|Histone H2B type 1 M|Histone H2B type 1 L|Histone protein|Histone H2B type 1 B|H2B.f|H2B.d|Histone H2B type 1 H|H2B histone family member O|H2BFF|H2BFD|HIRIP2|HIST1H2BH|H2B.c|H2BFS|H2B.1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Histone H2B
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fibre in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fibre is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H2B family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6p22-p21.3.
Type: Primary
Antigen: Histone H2B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-126/126
Host: Rabbit
Isotype: IgG
Reactivity: