Anti-Noggin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2975R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Noggin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:28-130/232
- Form:Liquid
- Gene ID:9241
- Antigen synonyms:SYM1|SYNS1|SYM 1|NOG|Symphalangism 1 proximal|Noggin|NOGG_HUMAN|SYNS 1|Synostoses multiple syndrome 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:23 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Noggin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Inhibitor of bone morphogenetic proteins (BMP) signaling which is required for growth and patterning of the neural tube and somite. Essential for cartilage morphogenesis and joint formation. Inhibits chondrocyte differentiation through its interaction with GDF5 (PubMed:21976273).
Type: Primary
Antigen: Noggin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 28-130/232
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Others