Anti-IL-18R Beta/CD218b Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2616R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IL-18R Beta/CD218b
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:15-120/599
- Form:Liquid
- Gene ID:8807
- Antigen synonyms:IL-18Rbeta|IL-1RAcPL|CDw218b|Interleukin-18 receptor beta|CD218 antigen-like family member B|IL-1R accessory protein-like|IL18RAP|Interleukin-18 receptor accessory protein|IL-18 receptor accessory protein|Interleukin-1 receptor 7|CD218b|IL18RB|Accessory protein-like|IL-1R-7|IL-1R7|IL-18R-beta|ACPL|Interleukin-18 receptor accessory protein-like|IL-18RAcP|IL1R7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:68 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IL-18R beta
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Required for the high affinity binding of interleukin 18 (IL-18) to its receptor complex (By similarity). Together with IL18R1 mediates IL-18-dependent activation of NF-kappa-B and JNK.
Type: Primary
Antigen: IL-18R Beta/CD218b
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 15-120/599
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse